Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTCCCTTCGGACACTCAGCGT[C/T]CTAGGGAGTCCCTTTGCAGAGTCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh22 PubMed Links |
Cdh22 - cadherin 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174988.3 | Intron | NP_778153.2 |