Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTATTGTTTACCTGTTTTCTAC[C/T]GCTATTAAGAGTTCCTGGCACAAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Strbp PubMed Links |
Strbp - spermatid perinuclear RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009261.3 | Intron | NP_033287.2 |