Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTACACAGCCACCTTGCTAGC[C/T]CCATATGCTCCCACAATCAGGTCTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Itga2b PubMed Links |
Itga2b - integrin alpha 2b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010575.2 | 1584 | Silent Mutation | NP_034705.2 |