Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCGAGGGCTGCAGGAAGTGAGGA[C/T]GATACTGGGGGGTTGGGTGGGAACG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pkp1 PubMed Links |
Pkp1 - plakophilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019645.2 | 3941 | UTR 3 | NP_062619.1 |