Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAAAAGACTCTGGATGAATTGA[C/T]GCTCAAAGCAACTGCACATGTTAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Srpk2 PubMed Links |
Srpk2 - serine/arginine-rich protein specific kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009274.2 | Intron | NP_033300.2 |