Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGTATTCAGCAACAGCGGTCA[C/T]GTCACCCAAGACCCATATGACCTTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Megf10 PubMed Links |
Megf10 - multiple EGF-like-domains 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001979.2 | 3937 | Silent Mutation | NP_001001979.1 |