Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGAGGGACATACCTGATGGTAG[A/G]GCCTGACACCTGACGCCCAGAAGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2310039H08Rik PubMed Links |
2310039H08Rik - RIKEN cDNA 2310039H08 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Ptcra - pre T cell antigen receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011195.2 | 167 | Silent Mutation | NP_035325.1 |