Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATAATATATTTTGAAATGATGT[C/T]ACAACTTTCTTTTGAAGGCAGGAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pkp4 PubMed Links |
Pkp4 - plakophilin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026361.2 | Intron | NP_080637.1 | ||||
NM_175464.2 | Intron | NP_780673.2 |