Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGCTTTATTTTTCCTCGTGGAT[A/G]TTCTTCTTAGAGTGTCTGTAGAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tpte PubMed Links |
Tpte - transmembrane phosphatase with tensin homology | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199257.2 | 1158 | Missense Mutation | NP_954866.2 |