Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCAGCTATCATTCTTTGGAGCAC[A/G]GCATCTTGGAAACAAGTGCAGAGTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Elp2 PubMed Links |
Elp2 - elongator acetyltransferase complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021448.2 | 1837 | Silent Mutation | NP_067423.2 |