Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTGGCACTTAAAGCATTGGGA[A/G]GTTGCTATAATAATCTAGAAGCCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mgat5 PubMed Links |
Mgat5 - mannoside acetylglucosaminyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145128.3 | Intron | NP_660110.2 |