Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAGAACCACATAAAGATCCTTT[C/T]CAGAGTCTCCCTGCTGCCCCATCCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ebf2 PubMed Links |
Ebf2 - early B cell factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276387.1 | Intron | NP_001263316.1 | ||||
NM_010095.6 | Intron | NP_034225.1 |