Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGATAAGGGGACACCCTACAA[A/G]TTCCACAACATATCTGGAATGGGTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Myoz2 PubMed Links |
Myoz2 - myozenin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021503.2 | Intron | NP_067478.1 |