Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGCTGACCTTCTGAAGCAGGA[A/C]TCATTCCCTCCCTCCAGTTGCTTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Eya4 PubMed Links |
Eya4 - eyes absent 4 homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010167.4 | Intron | NP_034297.2 |