Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCAAGACTACATTAAAAAAAA[A/T]TTATGGCCATCCAGAACTACAACTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sema6a PubMed Links |
Sema6a - sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018744.2 | Intron | NP_061214.2 |