Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCATGTTGAGGTTAAACCATC[A/G]AAACCATGTGATGCCAACAAACAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptprk PubMed Links |
Ptprk - protein tyrosine phosphatase, receptor type, K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008983.2 | Intron | NP_033009.1 |