Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGCCTGACTCGACTGAGGAGGC[A/G]GATATAGCTGCAGTGTAGCTTGCTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Klhl29 PubMed Links |
Klhl29 - kelch-like 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164493.1 | Intron | NP_001157965.1 |