Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGTGGATTTTAACTAGGAATTA[C/T]AAAGCCCTCAGCAAAGGTTCCAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Chd1 PubMed Links |
Chd1 - chromodomain helicase DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007690.3 | 2689 | Silent Mutation | NP_031716.2 |