Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTCTGCTTAGTTGAGCTTTATA[A/G]TATACCCAGCAGCCTTGACCATAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Myh10 PubMed Links |
Myh10 - myosin, heavy polypeptide 10, non-muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175260.2 | Intron | NP_780469.1 |