Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACAACCCATTGCTTGAGCCTGAG[C/T]CACTCAGCTAAGGAGAGCCTATGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tmprss7 PubMed Links |
Tmprss7 - transmembrane serine protease 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172455.3 | Intron | NP_766043.3 |