Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTGTGTACAATACTTACTATA[C/T]GAGTAGTCAACCTCTGCTAATATGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zfhx4 PubMed Links |
Zfhx4 - zinc finger homeodomain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030708.2 | Intron | NP_109633.2 |