Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Atp1b2 PubMed Links |
Atp1b2 - ATPase, Na+/K+ transporting, beta 2 polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013415.5 | Intron | NP_038201.1 |
Shbg - sex hormone binding globulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |