Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCTATGAACTAGTCATAGCCTG[A/G]TTATTGAATCCAAGAGAGATATGTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sulf1 PubMed Links |
Sulf1 - sulfatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198565.1 | Intron | NP_001185494.1 | ||||
NM_001198566.1 | Intron | NP_001185495.1 | ||||
NM_172294.1 | Intron | NP_758498.1 |