Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGCGTTGTCATCACCATCATC[C/T]GTGTCCTCTAAATCCATATCCACTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ubr5 PubMed Links |
Ubr5 - ubiquitin protein ligase E3 component n-recognin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081359.3 | 7911 | Silent Mutation | NP_001074828.2 | |||
NM_001112721.2 | 7911 | Silent Mutation | NP_001106192.1 |