Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATGAAGAATAATTTCTTCAAAG[A/G]ATCGTGGGTCCGTTCTGTTTATAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zbtb20 PubMed Links |
Zbtb20 - zinc finger and BTB domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285805.1 | Intron | NP_001272734.1 | ||||
NM_019778.2 | Intron | NP_062752.2 | ||||
NM_181058.1 | Intron | NP_851401.1 |