Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAAAAGGTAAATTCCCATGATT[C/T]ACACAGGTATTAGCAGTTGAACTGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Snd1 PubMed Links |
Snd1 - staphylococcal nuclease and tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019776.2 | Intron | NP_062750.2 |