Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGAGTACAGCTCACCAAGGCCTC[C/T]TGGGCTTGCGCAGGAATCCAGAGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Urb1 PubMed Links |
Urb1 - URB1 ribosome biogenesis 1 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029497.1 | 2683 | Silent Mutation | NP_083773.1 |