Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTTTTAGGGGACAGATTCTGA[C/T]TTTCTGATCTTTATTTTCTGGGCTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tox PubMed Links |
Tox - thymocyte selection-associated high mobility group box | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145711.4 | Intron | NP_663757.3 |