Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGATGGTCAGACTTGTTACGTCC[C/T]TTCTCCCGTGGTCATATTCAGAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Map4k4 PubMed Links |
Map4k4 - mitogen-activated protein kinase kinase kinase kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252200.1 | Intron | NP_001239129.1 | ||||
NM_001252201.1 | Intron | NP_001239130.1 | ||||
NM_001252202.1 | Intron | NP_001239131.1 | ||||
NM_008696.2 | Intron | NP_032722.2 |