Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCTTGAGGGCCGGAAGAAACAA[C/T]AGTTATTCATTCAGTACTCATAAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rbbp5 PubMed Links |
Rbbp5 - retinoblastoma binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172517.2 | Intron | NP_766105.2 |