Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTACTGATGTACACAAACATCC[A/G]TTTCTCTTATGGGAGACAAACAATC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Daam2 PubMed Links |
Daam2 - dishevelled associated activator of morphogenesis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008231.2 | Intron | NP_001008232.2 |