Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATGTAGCAAACAACAAAACCAT[C/T]AGTTGTGCTATTTGGTCCTTTAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cul3 PubMed Links |
Cul3 - cullin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016716.4 | Intron | NP_057925.1 |