Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGGCAACAACGCCACTTACCA[C/T]GGAACTAAAGGAGAACACACCTCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ctnnd2 PubMed Links |
Ctnnd2 - catenin (cadherin associated protein), delta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008729.2 | 3897 | Silent Mutation | NP_032755.2 |