Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGATGGCAACCCTGTCCCCAGTGA[C/T]GGAAGTTCTGATGAGCAGCCCATGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ercc6l2 PubMed Links |
Ercc6l2 - excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013608.2 | 2567 | Silent Mutation | NP_001013626.2 | |||
NM_023507.3 | 2567 | Intron | NP_075996.2 |