Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTTGACATATTTACGCATAAGA[G/T]CTCAGGAATTGAAGGGTGCTTCTTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Epha6 PubMed Links |
Epha6 - Eph receptor A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007938.2 | Intron | NP_031964.2 |