Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCCTCTTTTTTAGGTTTACAGA[A/T]TTGTAACCAAGTAACAAATTGTAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slit3 PubMed Links |
Slit3 - slit homolog 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011412.3 | Intron | NP_035542.2 |