Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATTTGGACCTGAACTGTACCTC[A/G]ATGAAGAATGACATTAATAGGCAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ttll6 PubMed Links |
Ttll6 - tubulin tyrosine ligase-like family, member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172799.4 | 2133 | UTR 3 | NP_766387.2 |