Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTATAAAAGTATCAGTCAGCTTT[C/T]TCTTTGTGATATACAATGTAAGTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rasal2 PubMed Links |
Rasal2 - RAS protein activator like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177644.5 | Intron | NP_808312.3 |