Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGTCGACAACACCGTCTCTATCT[G/T]ATTTCTGGTGAGGAGCTGTTCATAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Krt222 PubMed Links |
Krt222 - keratin 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172946.2 | 93 | Missense Mutation | NP_766534.1 |