Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGACTTCCATGTGGATCGACCT[A/C]TTGAGGAGTCACTGTTGAACCCAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700015C15Rik PubMed Links |
1700015C15Rik - RIKEN cDNA 1700015C15 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Cdk20 - cyclin-dependent kinase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053180.2 | 1028 | Missense Mutation | NP_444410.1 |