Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGGCTTGGAGATGAACTACTG[C/T]AGGAACCCGGATGGTGACAAGGGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Plg PubMed Links |
Plg - plasminogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008877.3 | 1337 | Silent Mutation | NP_032903.3 |