Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATTGAAGAAACAAGCTCTACTT[A/C]ACCTTTCAAACAACAGGCAGTGAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ctso PubMed Links |
Ctso - cathepsin O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177662.2 | Intron | NP_808330.1 |