Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCGCTGTACCATTTTATGCCAAC[A/T]CATTCTCTATTCCTCTTGTGGCTTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hmcn1 PubMed Links |
Hmcn1 - hemicentin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024720.3 | Intron | NP_001019891.2 |