Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTCTGTGCTATATTCAATTGG[C/T]TATAAGACCTGGAACTTAGTATGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Prkce PubMed Links |
Prkce - protein kinase C, epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011104.3 | Intron | NP_035234.1 |