Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAATGGGAGGCACCTGATGGAT[C/T]TGATCTGCTTAAACTCGGCCAGGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptk2b PubMed Links |
Ptk2b - PTK2 protein tyrosine kinase 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162365.1 | 1124 | Missense Mutation | NP_001155837.1 | |||
NM_001162366.1 | 1124 | Missense Mutation | NP_001155838.1 | |||
NM_172498.3 | 1124 | Missense Mutation | NP_766086.2 |