Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGGGAGGCTGCTGCTCTGCT[C/T]CGACTGTTTTCTCAATGAATAGCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sema3e PubMed Links |
Sema3e - sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011348.2 | 281 | UTR 5 | NP_035478.2 |