Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCAGGAGGTGAGCGAACCCCT[C/T]ACAGCCACCAAGCACCATGGCGGCG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptpn14 PubMed Links |
Ptpn14 - protein tyrosine phosphatase, non-receptor type 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008976.2 | 2420 | Silent Mutation | NP_033002.2 |