Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGCCTGCTCTGGCCATATTCC[A/C]GAGTCCATGACAATCCTTCCCCCTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Grin3a PubMed Links |
Grin3a - glutamate receptor ionotropic, NMDA3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033351.2 | 2167 | Silent Mutation | NP_001028523.1 | |||
NM_001276355.1 | 2167 | Silent Mutation | NP_001263284.1 |