Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTACTGATGAGGTTGGTTGAGA[C/G]CCAGAATGGGAAGGGAAGGGGAATC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Chst11 PubMed Links |
Chst11 - carbohydrate sulfotransferase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021439.2 | Intron | NP_067414.2 |