Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAAGTAGTGCAAAGATACGGGG[A/G]CCTGGGCACTCCTGTGGAAGAGCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pcnxl4 PubMed Links |
Pcnxl4 - pecanex-like 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026327.3 | Intron | NP_080603.2 |